Learn More
2-Propenyl-(propyl-d7)-propanedioic Acid Diethyl Ester, TRC
Product Code. 30427188
Change view
Click to view available options
Kvantitet:
25 mg
Product Code. Kvantitet
30427188 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30427188 Supplier Toronto Research Chemicals Supplier No. TRCP76900225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2-Propenyl-(propyl-d7)-propanedioic Acid Diethyl Ester
CAS 1215738-07-0
Molekylformel C13H15D7O4
Kvantitet 25 mg
InChI formel InChI=1S/C13H22O4/c1-5-9-13(10-6-2,11(14)16-7-3)12(15)17-8-4/h5H,1,6-10H2,2-4H3/i2D3,6D2,10D2
LEDER [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C(CC=C)(C(=O)OCC)C(=O)OCC
IUPAC-namn diethyl 2-(1,1,2,2,3,3,3-heptadeuteriopropyl)-2-prop-2-enylpropanedioate
Molekylvikt (g/mol) 249.35
Formel vikt 249.1957