Läs mer
2-Propyl-2-heptenal (Technical Grade), TRC
Produktkod. 30423919
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30423919 10 g
30428520 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423919 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP83581510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34880-43-8
Molekylformel C10H18O
Molekylvikt (g/mol) 154.25
Synonym 2-Propyl-2-hepten-1-al
LEDER CCCCCC(CCC)C=O
Specifikationer
Kemiskt namn eller material 2-Propyl-2-heptenal (Technical Grade)
Kvantitet 10 g
InChI formel InChI=1S/C10H18O/c1-3-5-6-8-10(9-11)7-4-2/h8-9H,3-7H2,1-2H3/b10-8+