Learn More
2-Propylglutaric Acid, TRC
Product Code. 30417827
Change view
Click to view available options
Kvantitet:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
30417827 5 mg -
30421680 2.5 mg 2.5mg
30428411 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30417827 Supplier Toronto Research Chemicals Supplier No. TRCP8353005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32806-62-5
Molekylformel C8 H14 O4
Molekylvikt (g/mol) 174.19
IUPAC-namn 2-propylpentanedioic acid
LEDER CCCC(CCC(=O)O)C(=O)O
Specifications
Kemiskt namn eller material 2-Propylglutaric Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C8H14O4/c1-2-3-6(8(11)12)4-5-7(9)10/h6H,2-5H2,1H3,(H,9,10)(H,11,12)
Formel vikt 174.0892
Procent renhet >95%