Läs mer
2-Propylhexanoic Acid Methyl Ester, TRC
Produktkod. 30410163
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30410163 500 mg
1 options
Produktkod. 30410163 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP837105500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Propylhexanoic Acid Methyl Ester
CAS 5162-60-7
Molekylformel C10H20O2
Kvantitet 500 mg
Synonym 4-Octanecarboxylic Acid Methyl Ester
InChI formel InChI=1S/C10H20O2/c1-4-6-8-9(7-5-2)10(11)12-3/h9H,4-8H2,1-3H3
LEDER O=C(OC)C(CCC)CCCC
IUPAC-namn methyl 2-propylhexanoate
Molekylvikt (g/mol) 172.26
Formel vikt 172.1463