Läs mer
2-Propyloxybenzamidine Hydrochloride, TRC
Produktkod. 30421125
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30421125 100 mg
1 options
Produktkod. 30421125 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP835750100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Propyloxybenzamidine','Hydrochloride
CAS 57075-84-0
Molekylformel C10 H14 N2 O . Cl H
Kvantitet 100 mg
Synonym Benzenecarboximidamide','2-propoxy-','hydrochloride (1:1), Benzenecarboximidamide','2-propoxy-','monohydrochloride (9CI), 2-Propoxybenzamidine hydrochloride, 2-Propyloxybenzamidine hydrochloride, 2-Propoxybenzene-1-carboximidamide hydrochloride, 2-Propoxybenzenecarboximidamide hydrochloride, 2-Propyloxybenzimidamide hydrochloride
InChI formel InChI=1S/C10H14N2O.ClH/c1-2-7-13-9-6-4-3-5-8(9)10(11)12;/h3-6H,2,7H2,1H3,(H3,11,12);1H
LEDER Cl.CCCOc1ccccc1C(=N)N
IUPAC-namn 2-propoxybenzenecarboximidamide;hydrochloride
Molekylvikt (g/mol) 214.69
Formel vikt 214.0873