Läs mer
2-Propyltriphenylphosphonium Bromide, TRC
Produktkod. 30420673
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30420673 100 mg
1 options
Produktkod. 30420673 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP838335100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Propyltriphenylphosphonium Bromide
CAS 1530-33-2
Molekylformel C21H22BrP
Kvantitet 100 mg
Synonym Isopropyltriphenylphosphonium Bromide, (1-Methylethyl)triphenyl-phosphonium Bromide, Isopropyltriphenylphosphonium Bromide, (1-Methylethyl)triphenylphosphonium Bromide, NSC 113130
InChI formel InChI=1S/C21H22P.BrH/c1-18(2)22(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-18H,1-2H3;1H/q+1;/p-1
LEDER [Br-].CC(C)[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC-namn triphenyl(propan-2-yl)phosphanium;bromide
Molekylvikt (g/mol) 385.28
Formel vikt 384.0642