missing translation for 'onlineSavingsMsg'
Learn More

(2-Pyrid-3-ylphenyl)methanol, ≥97%, Thermo Scientific™

Catalog Number p-3176003
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
857284-03-8
C12H11NO
185.226
MFCD08060473
QREMDJYNQPJMFO-UHFFFAOYSA-N
2-pyrid-3-ylphenyl methanol, 2-pyridin-3-yl phenyl methanol, 2-pyridin-3-ylphenyl methanol, 2-pyridin-3-yl-phenyl-methanol, 2-3-pyridyl phenyl methan-1-ol, benzenemethanol, 2-3-pyridinyl
7537538
(2-pyridin-3-ylphenyl)methanol
C1=CC=C(C(=C1)CO)C2=CN=CC=C2
This item is not returnable. View return policy
857284-03-8
C12H11NO
185.226
MFCD08060473
QREMDJYNQPJMFO-UHFFFAOYSA-N
2-pyrid-3-ylphenyl methanol, 2-pyridin-3-yl phenyl methanol, 2-pyridin-3-ylphenyl methanol, 2-pyridin-3-yl-phenyl-methanol, 2-3-pyridyl phenyl methan-1-ol, benzenemethanol, 2-3-pyridinyl
7537538
(2-pyridin-3-ylphenyl)methanol
C1=CC=C(C(=C1)CO)C2=CN=CC=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 185.22
Percent Purity 97%
Chemical Name or Material (2-Pyrid-3-ylphenyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'