Läs mer
2-Pyridinesulfonyl Chloride, Technical Grade, TRC
Produktkod. 30417025
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30417025 500 mg
1 options
Produktkod. 30417025 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP991550500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Pyridinesulfonyl Chloride','Technical Grade
CAS 66715-65-9
Molekylformel C5H4ClNO2S
Kvantitet 500 mg
Synonym Pyridine-2-sulfonyl Chloride, 2-Chlorosulfonyl-pyridinium Chloride
InChI formel InChI=1S/C5H4ClNO2S/c6-10(8,9)5-3-1-2-4-7-5/h1-4H
LEDER C1=CC=NC(=C1)S(=O)(=O)Cl
IUPAC-namn pyridine-2-sulfonyl chloride
Molekylvikt (g/mol) 177.61
Formel vikt 176.97