missing translation for 'onlineSavingsMsg'
Learn More

2-(Pyrrolidin-1-yl)pyrimidine-5-boronic acid pinacol ester, 95%, ACROS Organics™

Brand:  Acros Organics 454350010

Additional Details : CAS Number : 1015242-07-5 Weight : 0.00100kg


Quantity 1g

 View more versions of this product

Product Code. 15204348

  • 5605.61 SEK / 1g


Estimated Shipment
04-11-2020
Add to basket


Specifications

Chemical Identifiers

1015242-07-5
275.16
2-pyrrolidin-1-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-pyrrolidin-1-yl pyrimidine-5-boronic acid pinacol ester, 2-pyrrolidinopyrimidine-5-boronic acid pinacol ester, 2-pyrrolidin-1-yl pyrimidin-5-ylboronic acid pinacol ester, 2-pyrrolidin-1-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-pyrrolidinopyrimidine-5-boronic acid, pinacol ester, 2-pyrrolidin-1-yl pyrimidine-5-boronic acid, pinacol ester, 2-pyrrolidin-1-yl pyrimidin-5-yl boronic acid pinacol ester
2-pyrrolidin-1-yl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
C14H22BN3O2
CGSCNCRVBHIXSO-UHFFFAOYSA-N
16414224
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)N3CCCC3

Specifications

2-(Pyrrolidin-1-yl)pyrimidine-5-boronic acid pinacol ester
Glass Bottle
1g
94% min. (GC)
2-pyrrolidin-1-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-pyrrolidin-1-yl pyrimidine-5-boronic acid pinacol ester, 2-pyrrolidinopyrimidine-5-boronic acid pinacol ester, 2-pyrrolidin-1-yl pyrimidin-5-ylboronic acid pinacol ester, 2-pyrrolidin-1-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-pyrrolidinopyrimidine-5-boronic acid, pinacol ester, 2-pyrrolidin-1-yl pyrimidine-5-boronic acid, pinacol ester, 2-pyrrolidin-1-yl pyrimidin-5-yl boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)N3CCCC3
275.16
275.16
Authentic
151.0°C to 152.0°C
1015242-07-5
C14H22BN3O2
CGSCNCRVBHIXSO-UHFFFAOYSA-N
2-pyrrolidin-1-yl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
16414224
95%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-(Pyrrolidin-1-yl)pyrimidine-5-boronic acid pinacol ester

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documents