Läs mer
2-Pyrrolidinyl Desmorpholinylrocuronium Bromide, TRC
Produktkod. 30417318
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417318 10 mg
30427451 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30417318 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99854010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1190105-65-7
Molekylformel C32 H53 N2 O3 . Br
Molekylvikt (g/mol) 593.68
Synonym Pyrrolidinium, 1-[(2β,3α,5α,16β,17β)-17-(acetyloxy)-3-hydroxy-2-(1-pyrrolidinyl)androstan-16-yl]-1-(2-propen-1-yl)-, bromide (1:1), 1-[17β-Acetoxy-3α-hydroxy-2β-(pyrrolidin-1-yl)-5α-androstan-16β-yl]-1-(prop-2-enyl)pyrrolidinium Bromide
IUPAC-namn [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-3-hydroxy-10,13-dimethyl-16-(1-prop-2-enylpyrrolidin-1-ium-1-yl)-2-pyrrolidin-1-yl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate;bromide
LEDER [Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N5CCCC5)[N+]6(CC=C)CCCC6
Specifikationer
Kemiskt namn eller material 2-Pyrrolidinyl Desmorpholinylrocuronium bromide
Kvantitet 10 mg
InChI formel InChI=1S/C32H53N2O3.BrH/c1-5-16-34(17-8-9-18-34)28-20-26-24-11-10-23-19-29(36)27(33-14-6-7-15-33)21-32(23,4)25(24)12-13-31(26,3)30(28)37-22(2)35;/h5,23-30,36H,1,6-21H2,2-4H3;1H/q+1;/p-1/t23-,24+,25-,26-,27-,28-,29-,30-,31-,32-;/m0./s1
Formel vikt 592.324