Läs mer
2-Pyrrolidinylphosphonic Acid, TRC
Produktkod. 30425613
Change view
Kvantitet:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425613 10 mg
30408995 100 mg
30426461 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425613 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99185510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73858-59-0
Molekylformel C4 H10 N O3 P
Molekylvikt (g/mol) 151.1
Synonym Phosphonic acid, 2-pyrrolidinyl- (9CI), Pyrrolidin-2-ylphosphonic Acid
IUPAC-namn pyrrolidin-2-ylphosphonic acid
LEDER OP(=O)(O)C1CCCN1
Specifikationer
Kemiskt namn eller material 2-Pyrrolidinylphosphonic acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C4H10NO3P/c6-9(7,8)4-2-1-3-5-4/h4-5H,1-3H2,(H2,6,7,8)
Formel vikt 151.0398
Procent renhet >95%