Läs mer
(2'S)-Nicotine 1-Oxide, TRC
Produktkod. 18000450
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18000450 100 mg
18062950 50 mg
18042050 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18000450 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN428000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2820-55-5
Molekylformel C10 H14 N2 O
Molekylvikt (g/mol) 178.23
Synonym Pyridine, 3-[(2S)-1-methyl-2-pyrrolidinyl]-, 1-oxide, Nicotine N-oxide (6CI), Nicotine, 1-oxide (7CI,8CI), Pyridine, 3-(1-methyl-2-pyrrolidinyl)-, 1-oxide, (S)-, 3-[(2S)-1-Methylpyrrolidin-2-yl]pyridin-1-ium-1-olate, Nicotine 1-N-oxide, 3-[(2S)-1-Methylpyrrolidin-2-yl]pyridine 1-oxide, (2S)-1-Methyl-2-(1-oxido-3-pyridinyl)pyrrolidine, (2'S)-Nicotine 1-oxide
IUPAC-namn 3-[(2S)-1-methylpyrrolidin-2-yl]-1-oxidopyridin-1-ium
LEDER CN1CCC[C@H]1c2ccc[n+]([O-])c2
Specifikationer
Kemiskt namn eller material (2'S)-Nicotine 1-Oxide
Anteckningar om renhetsgrad GC
Kvantitet 100 mg
InChI formel InChI=1S/C10H14N2O/c1-11-6-3-5-10(11)9-4-2-7-12(13)8-9/h2,4,7-8,10H,3,5-6H2,1H3/t10-/m0/s1
Formel vikt 178.1106
Procent renhet >95%