Läs mer
2-tert-Butoxy-3-iodopyridine, 95%, Thermo Scientific™
Produktkod. 11456560
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11456560 1 g 1g
11466560 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11456560 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H27620.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1020253-78-4
Molekylformel C9H12INO
Molekylvikt (g/mol) 277.105
MDL-nummer MFCD09263324
InChI-nyckel BTQSPOKSJDXPLC-UHFFFAOYSA-N
Synonym 2-tert-butoxy-3-iodopyridine
PubChem CID 42553053
IUPAC-namn 3-iodo-2-[(2-methylpropan-2-yl)oxy]pyridine
LEDER CC(C)(C)OC1=C(C=CC=N1)I
Specifikationer
Kemiskt namn eller material 2-tert-Butoxy-3-iodopyridine
Kvantitet 1 g
Känslighet Light sensitive
Formel vikt 277.11
Procent renhet 95%