Learn More
[2-(tert-Butoxy)ethyl](methyl)amine, TRC
Product Code. 30344686
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30344686 50 mg 50mg
1 options
Product Code. 30344686 Supplier Toronto Research Chemicals Supplier No. TRCB41646350MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material [2-(tert-Butoxy)ethyl](methyl)amine
CAS 1008119-69-4
Molecular Formula C7H17NO
Quantity 50 mg
InChI Formula InChI=1S/C7H17NO/c1-7(2,3)9-6-5-8-4/h8H,5-6H2,1-4H3
SMILES CNCCOC(C)(C)C
IUPAC Name N-methyl-2-[(2-methylpropan-2-yl)oxy]ethanamine
Molecular Weight (g/mol) 131.22
Formula Weight 131.131