missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-4-methylthiazole-5-methanol, 97%, Thermo Scientific™

Catalog Number 15455047
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1256483-61-0
C9H15NOS
185.29
MFCD16660039
HJESPMQKUUFUGH-UHFFFAOYSA-N
2-tert-butyl-4-methylthiazole-5-methanol, 2-tert-butyl-4-methyl-1,3-thiazol-5-yl methanol, 2-tert-butyl-5-hydroxymethyl-4-methylthiazole
73012579
(2-tert-butyl-4-methyl-1,3-thiazol-5-yl)methanol
CC1=C(CO)SC(=N1)C(C)(C)C
This item is not returnable. View return policy
1256483-61-0
C9H15NOS
185.29
MFCD16660039
HJESPMQKUUFUGH-UHFFFAOYSA-N
2-tert-butyl-4-methylthiazole-5-methanol, 2-tert-butyl-4-methyl-1,3-thiazol-5-yl methanol, 2-tert-butyl-5-hydroxymethyl-4-methylthiazole
73012579
(2-tert-butyl-4-methyl-1,3-thiazol-5-yl)methanol
CC1=C(CO)SC(=N1)C(C)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.085
Boiling Point 110°C (0.75 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.525
Quantity 250 mg
Formula Weight 185.29
Percent Purity 97%
Chemical Name or Material 2-tert-Butyl-4-methylthiazole-5-methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'