Läs mer
2-Tetradecylcyclobutanone, TRC
Produktkod. 30426345
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426345 100 mg
30423070 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30426345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT291540100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35493-47-1
Molekylformel C18 H34 O
Molekylvikt (g/mol) 266.46
Synonym Cyclobutanone, 2-tetradecyl-, 2-(n-Tetradecyl)cyclobutanone, 2-Tetradecylcyclobutanone
IUPAC-namn 2-tetradecylcyclobutan-1-one
LEDER CCCCCCCCCCCCCCC1CCC1=O
Specifikationer
Kemiskt namn eller material 2-Tetradecylcyclobutanone
Kvantitet 100 mg
InChI formel InChI=1S/C18H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-17-15-16-18(17)19/h17H,2-16H2,1H3
Formel vikt 266.261