Läs mer
2-Thiopheneethanol Tosylate, TRC
Produktkod. 30449529
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30449529 1 g
1 options
Produktkod. 30449529 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3448601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Thiopheneethanol Tosylate
CAS 40412-06-4
Molekylformel C13 H14 O3 S2
Kvantitet 1 g
Synonym 2-Thiopheneethanol 2-(4-Methylbenzenesulfonate), 2-(2-Thienyl)ethyl Toluene-p-sulfonate, 2-(2-Thienyl)ethyl Tosylate, 2-(2-Thiophene)ethanol Tosylate, 2-(Thien-2-yl)ethyl 4-Methylbenzenesulfonate, 2-Thiophene Ethanol Tosylate, 2-Thiopheneethanol p-Toluenesulfonate
InChI formel InChI=1S/C13H14O3S2/c1-11-4-6-13(7-5-11)18(14,15)16-9-8-12-3-2-10-17-12/h2-7,10H,8-9H2,1H3
LEDER Cc1ccc(cc1)S(=O)(=O)OCCc2cccs2
IUPAC-namn 2-thiophen-2-ylethyl 4-methylbenzenesulfonate
Molekylvikt (g/mol) 282.38
Formel vikt 282.0384