Läs mer
2-Thioxo-3-thiazolidinecarboxylic Acid Benzyl Ester, TRC
Produktkod. 30456620
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30456620 10 g
1 options
Produktkod. 30456620 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT39501010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Thioxo-3-thiazolidinecarboxylic Acid Benzyl Ester
CAS 74058-68-7
Molekylformel C11H11NO2S2
Kvantitet 10 g
Synonym 2-Thioxo-3-thiazolidinecarboxylic Acid Phenylmethyl Ester
InChI formel InChI=1S/C11H11NO2S2/c13-10(12-6-7-16-11(12)15)14-8-9-4-2-1-3-5-9/h1-5H,6-8H2
LEDER O=C(OCc1ccccc1)N2CCSC2=S
IUPAC-namn benzyl 2-sulfanylidene-1,3-thiazolidine-3-carboxylate
Molekylvikt (g/mol) 253.34
Formel vikt 253.0231