Läs mer
2-Thioxothiazolidine-4-carboxylic Acid, TRC
Produktkod. 30450117
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30450117 50 mg
30451123 25 mg
30456070 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30450117 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT36883050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20933-67-9
Molekylformel C4 H5 N O2 S2
Molekylvikt (g/mol) 163.22
Synonym 4-Thiazolidinecarboxylic acid, 2-thioxo-, 2-Thioxo-4-thiazolidinecarboxylic acid, TTCA, TTCA (thiram metabolite), Thiazolidine-2-thione-4-carboxylic acid
IUPAC-namn 2-sulfanylidene-1,3-thiazolidine-4-carboxylic acid
LEDER OC(=O)C1CSC(=S)N1
Specifikationer
Kemiskt namn eller material 2-Thioxothiazolidine-4-carboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C4H5NO2S2/c6-3(7)2-1-9-4(8)5-2/h2H,1H2,(H,5,8)(H,6,7)
Formel vikt 162.9762
Procent renhet >95%