Learn More
2-(Trifluoromethyl)benzene-1,4-diamine, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 364-13-6 |
| Molecular Formula | C7H7F3N2 |
| MDL Number | MFCD00036101 |
| Quantity | 100 g |
| SMILES | NC1=CC=C(N)C=C1C(F)(F)F |
| Molecular Weight (g/mol) | 176.14 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-(Trifluoromethyl)benzene-1,4-diamine |