missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7021679
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
776-04-5
C7H4ClF3O2S
244.612
MFCD00051696
ZIZGWNOAHUCACM-UHFFFAOYSA-N
2-trifluoromethyl benzenesulfonyl chloride, 2-trifluoromethyl benzene-1-sulfonyl chloride, 2-trifluoromethylbenzenesulphonyl chloride, 2-trifluoromethyl benzenesulphonyl chloride, o-trifluoromethyl benzenesulfonyl chloride, o-trifluoromethylbenzenesulfonyl chloride, 2-trifluoromethyl benzene sulfonyl chloride, benzenesulfonyl chloride, 2-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-2-sulfonyl chloride
136616
2-(trifluoromethyl)benzenesulfonyl chloride
C1=CC=C(C(=C1)C(F)(F)F)S(=O)(=O)Cl
This item is not returnable. View return policy
776-04-5
C7H4ClF3O2S
244.612
MFCD00051696
ZIZGWNOAHUCACM-UHFFFAOYSA-N
2-trifluoromethyl benzenesulfonyl chloride, 2-trifluoromethyl benzene-1-sulfonyl chloride, 2-trifluoromethylbenzenesulphonyl chloride, 2-trifluoromethyl benzenesulphonyl chloride, o-trifluoromethyl benzenesulfonyl chloride, o-trifluoromethylbenzenesulfonyl chloride, 2-trifluoromethyl benzene sulfonyl chloride, benzenesulfonyl chloride, 2-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-2-sulfonyl chloride
136616
2-(trifluoromethyl)benzenesulfonyl chloride
C1=CC=C(C(=C1)C(F)(F)F)S(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 22°C to 24°C
Density 1.585
Boiling Point 133°C to 135°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.503
Quantity 1 g
UN Number UN3261
Beilstein 2651854
Sensitivity Moisture sensitive
Formula Weight 244.62
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)benzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'