missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7045559
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
2285-12-3
C8H4F3NO
187.12
MFCD00002008
GZWGTVZRRFPVAS-UHFFFAOYSA-N
2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
16794
1-isocyanato-2-(trifluoromethyl)benzene
C1=CC=C(C(=C1)C(F)(F)F)N=C=O
This item is not returnable. View return policy
2285-12-3
C8H4F3NO
187.12
MFCD00002008
GZWGTVZRRFPVAS-UHFFFAOYSA-N
2-trifluoromethyl phenyl isocyanate, 1-isocyanato-2-trifluoromethyl benzene, benzene, 1-isocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isocyanate, o-trifluoromethylphenylisocyanate, isocyanic acid, alpha,alpha,alpha-trifluoro-o-tolyl ester, o-trifluoromethyl phenyl isocyanate, 2-trifluoromethyl phenylisocyanate, 2-trifluoromethyl benzenisocyanate, isocyanic acid, .alpha.,.alpha.,.alpha.-trifluoro-o-tolyl ester
16794
1-isocyanato-2-(trifluoromethyl)benzene
C1=CC=C(C(=C1)C(F)(F)F)N=C=O

2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)phenyl isocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Hydrolyzes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Density 1.238
Boiling Point 74°C to 75°C (40 mmHg)
Flash Point 58°C (136°F)
Odor Pungent
Refractive Index 1.4755
Quantity 1 g
UN Number UN2285
Beilstein 977056
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 187.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)phenyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'