Läs mer
2-Vinylthiophene (Stabilized with BHT), TRC
Produktkod. 30454193
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454193 50 mg
30455741 100 mg
30449044 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30454193 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV75699850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1918-82-7
Molekylformel C6H6S
Molekylvikt (g/mol) 110.17
LEDER C=Cc1cccs1
Specifikationer
Kemiskt namn eller material 2-Vinylthiophene
Kvantitet 50 mg
InChI formel InChI=1S/C6H6S/c1-2-6-4-3-5-7-6/h2-5H,1H2