Learn More
Vitlab™ 2-way Tube Connectors
Join tubes with varying diameters. Vitlab™ 2-way Tube connectors are perfect for linking tubes with variable diameters. The connectors can be attached to the next size up.
Brand: Vitlab™ 80436
Additional Details : Weight : 0.07200kg
Description
- Connect tubing of varying diameters
- For tubing with an inner diameter between 5-7mm
- Outlet opening 2,7mm
Specifications
7 to 9 mm | |
3.6 mm | |
20/Pk. |