Learn More
Vitlab™ 2-way Tube Connectors
Join tubes with varying diameters. Vitlab™ 2-way Tube connectors are perfect for linking tubes with variable diameters. The connectors can be attached to the next size up.
372.00 SEK - 565.00 SEK
Specifications
Packaging Quantity 20/Pk.
View More Specs
Products 5
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity & Availability  
15262749
View Documents Promotion Details
Vitlab™
80435
5-7 mm 2.7 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15272749
View Documents Promotion Details
Vitlab™
80436
7 to 9 mm 3.6 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15222779
View Documents Promotion Details
Vitlab™
80535
9 to 12 mm 3.6 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15232779
View Documents Promotion Details
Vitlab™
80536
11 to 14 mm 7.3 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15242779
View Documents Promotion Details
Vitlab™
80537
13 to 16 mm 8.8 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Connect tubing of varying diameters
  • For tubing with an inner diameter between 5-7mm
  • Outlet opening 2,7mm
Specifications
Specifications
20/Pk.