missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ -20 to +110 Lo Tox™ Laboratory Immersion Thermometer

Lo Tox™ organic filling is specially chosen to achieve wide temperature span and provide the lowest practical levels of hazard.

322.00 SEK - 326.00 SEK

Specifications

Length (Metric) 305 mm
Material Glass
Thermometer Fill Blue Organic Liquid
Product Type Laboratory Immersion Thermometer
Max. Temperature (Metric) 110°C
View More Specs

Products 2
Product Code Brand Immersion Type Price Quantity & Availability  
Product Code Brand Immersion Type Price Quantity & Availability  
15442695
View Documents Promotion Details
Fisherbrand™
Total Immersion
322.00 SEK
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
15422695
View Documents Promotion Details
Fisherbrand™
Partial Immersion
326.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
 
Description

Description

  • Quality and legibility make these an ideal substitute for mercury thermometers
  • Fitted with Brannan non roll end cap which has ring to allow the thermometer to be suspended
Specifications

Specifications

305 mm
Blue Organic Liquid
110°C
-20°C to +110°C
0.5°
Glass
Laboratory Immersion Thermometer
-20°C
±1°C at 35°C
Videos
SDS
Documents

Documents

Product Certifications