Läs mer
TdB Labs AB 20 kDa Q-dextran for Ion-exchange Chromatography and Biomolecular Studies
Produktkod. 30216325
Encompass_Preferred
Change view
Kvantitet:
1 g
10 g
Förpackningsstorlek:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30216325 1 g 1g
30216524 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30216325 Leverantör TdB Labs AB Leverantörsnummer QD20_1g
 Begränsad artikel
Beräknad leverans: 06-08-2026
Lägg i varukorgen
Lägg i varukorgen
Q-dextran is a positively charged dextran modified with quaternary ammonium groups. It is used in ion exchange chromatography, biochemical assays, and for studying electrostatic interactions with negatively charged biomolecules.
  • Storage and Stability:
    • Store dry at room temperature, protected from moisture and light.
    • Stable for extended periods under recommended storage conditions without loss of charge properties.
  • Specifikationer
    Kvantitet 1 g
    Produkttyp Modified Dextran