Learn More
Eppendorf™ 2000μL Sterile Polypropylene 96-Well Deep Well PCR Plates
PCR plates, free of DNA, DNase, RNase and PCR inhibitors
Brand:  Eppendorf™ 0030509315
Additional Details : Weight : 9.33300kg
Product Code. 10035254
  • 10815.00 SEK / Pack of 80
Estimated Shipment: 23-05-2024
to see stock.
Description
Description
These plates combine a new well design with a sophisticated color coding system for ease of reading and orientation. The well geometry allows high speed pipetting without splashing, excellent mixing and high speed centrifugation, which accelerates your manual and automated work processes. The new polypropylene plates fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines.
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • Free from DNA, DNase, RNase and PCR* inhibitor (PCR* clean)
  • Autoclavable
  • Optionally available with barcode
Specifications
Specifications
8 x 12
Polypropylene
Sterile
Clear
96