Läs mer
Endecotts™ Brass and Stainless Steel Test Sieve, 200 mm dia.
Porstorlek:
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
1 mm
106 μm
1,18 mm
125 μm
1,4 mm
150 μm
1,7 mm
180 μm
2 mm
212 μm
2,36 mm
250 μm
2,8 mm
300 μm
3,35 mm
355 μm
4 mm
425 μm
4,75 mm
500 μm
5,6 mm
600 μm
6,7 mm
710 μm
8 mm
850 μm
9,5 mm
11,2 mm
16 mm
Förpackningsstorlek:
Varje
Beskrivning
Full height test sieves provide all types of laboratory sampling and particle size analysis. Endecotts™ Brass Frame Test Sieves comply with ISO 3310-1 / BS410 standards.
- Precision frame constructed of Brass (ensures consistent nestability)
- Stainless steel mesh
- Precise aperture in accordance with ISO 3310-1 / BS410
- Natural fillet (free flowing of sample)
- Totally sealed (no crevice to lose material)
- Evenly tensioned mesh (ensures accurate analysis)
- Safe edge (big radius makes it comfortable to handle)
- Serial number (ensures full traceability)
Specifikationer
Specifikationer
| Kvantitet | 1Each |
| Porstorlek | 850 μm |
| Typ | Test Sieve |
| Certifikat och efterlevnad | ISO 3310-1 / BS410 |
| Material | Brass |
| Höjd (metrisk) | Full |
| För användning med (applikation) | For all types of laboratory sampling and particle size analysis |
| Diameter (metrisk) Yttre | 200 mm |
| Rammaterial | Brass |
| Mesh material | Stainless Steel |