Läs mer
20beta-Hydroxy Prednisone, TRC
Produktkod. 30006335
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006335 5 mg
30005758 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006335 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9522955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 600-92-0
Molekylformel C21H28O5
Molekylvikt (g/mol) 360.44
Synonym Pregna-1,4-diene-3,11-dione, 17,20,21-trihydroxy-, (20R)-, Pregna-1,4-diene-3,11-dione, 17,20β,21-trihydroxy- (8CI), (20R)-17,20,21-Trihydroxypregna-1,4-diene-3,11-dione, 17α,20β,21-Trihydroxypregna-1,4-diene-3,11-dione, 20α-Dihydroprednisone, Pregna-1,4-dien-17α,20β,21-triol-3,11-dione, Pregna-1,4-diene-17α,20β,21-triol-3,11-dione
IUPAC-namn (8S,9S,10R,13S,14S,17R)-17-[(1R)-1,2-dihydroxyethyl]-17-hydroxy-10,13-dimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione
LEDER C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)[C@H](O)CO
Specifikationer
Kemiskt namn eller material 20-beta-Hydroxy Prednisone
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-15,17-18,22,25-26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,17+,18+,19-,20-,21-/m0/s1
Formel vikt 360.1937
Procent renhet >95%