Läs mer
21-Acetyl Beclomethasone, TRC
Produktkod. 30343984
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30343984 500 mg
1 options
Produktkod. 30343984 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA777130500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beclomethasone 21-acetate
CAS 4735-64-2
Molekylformel C24 H31 Cl O6
Kvantitet 500 mg
Synonym (11beta,16beta)-21-(Acetyloxy)-9-chloro-11,17-dihydroxy-16-methylpregna-1,4-diene-3,20-dione, Beclomethasone 21-acetate, (11β,16β)-21-(Acetyloxy)-9-chloro-11,17-dihydroxy-16-methylpregna-1,4-diene-3,20-dione (ACI), Pregna-1,4-diene-3,20-dione, 9-chloro-11β,17,21-trihydroxy-16β-methyl-, 21-acetate (7CI, 8CI)
InChI formel InChI=1S/C24H31ClO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17-,18-,19-,21-,22-,23-,24-/m0/s1
LEDER C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molekylvikt (g/mol) 450.952
Formel vikt 450.181