Läs mer
21-(Acetyloxy)-9-chloro-11beta-hydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate (Beclometasone 21-Acetate 17-Propionate), Mikromol™
Produktkod. 30463295
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30463295 4 x 25 mg
30049286 25 mg
17911276 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30463295 Leverantör Mikromol Leverantörsnummer MM0199.0600254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
21-(Acetyloxy)-9-chloro-11beta-hydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate (Beclometasone 21-Acetate 17-Propionate), Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Beclomethasone 21-Acetate 17-Propionate
CAS 8-7-5534
Analyt- eller komponentnamn Beclomethasone 21-Acetate 17-Propionate
Frakt skick Room Temperature
Molekylformel C27 H35 Cl O7
Certifikat och efterlevnad ISO 9001
Synonym Beclometasone Dipropionate Imp. B (EP), 21-(Acetyloxy)-9-chloro-11beta-hydroxy-16beta-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate, Beclometasone 21-Acetate 17-Propionate, Beclometasone Dipropionate Impurity B
InChI formel InChI=1S/C27H35ClO7/c1-6-23(33)35-27(22(32)14-34-16(3)29)15(2)11-20-19-8-7-17-12-18(30)9-10-24(17,4)26(19,28)21(31)13-25(20,27)5/h9-10,12,15,19-21,31H,6-8,11,13-14H2,1-5H3/t15-,19-,20-,21-,24-,25-,26-,27-/m0/s1
Lösningstyp Neat
LEDER CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C)C(=O)COC(=O)C
IUPAC-namn [(8S,9R,10S,11S,13S,14S,16S,17R)-17-(2-acetyloxyacetyl)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molekylvikt (g/mol) 507.02
Kvantitet 4 x 25 mg
Formel vikt 506.2071 g/mol