Läs mer
21-(Acetyloxy) Triamcinolone Acetonide, TRC
Produktkod. 30320346
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30320346 1 mg
1 options
Produktkod. 30320346 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1874201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triamcinolone Acetonide 21-Acetate
CAS 3870-07-3
Molekylformel C26 H33 F O7
Kvantitet 1 mg
Synonym (11beta,16alpha)-21-(acetyloxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione, 9-Fluoro-11β,16α,17,21-tetrahydroxy-pregna-1,4-diene-3,20-dione 21-acetate cyclic 16,17-acetal with acetone, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-diene-3,20-dione deriv. Triamcinolone acetonide 21-acetate
InChI formel InChI=1S/C26H33FO7/c1-14(28)32-13-20(31)26-21(33-22(2,3)34-26)11-18-17-7-6-15-10-16(29)8-9-23(15,4)25(17,27)19(30)12-24(18,26)5/h8-10,17-19,21,30H,6-7,11-13H2,1-5H3/t17-,18-,19-,21+,23-,24-,25-,26+/m0/s1
LEDER CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C
IUPAC-namn (11β,16α)-21-(Acetyloxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione
Molekylvikt (g/mol) 476.53
Formel vikt 476.221