Läs mer
21-Carboxy Dexamethasone, TRC
Produktkod. 17960705
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17960705 50 mg
17952905 10 mg
17903115 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17960705 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF45850050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84449-15-0
Molekylformel C22 H27 F O6
Molekylvikt (g/mol) 406.44
Synonym (11β,16α)-9-Fluoro-11,17-dihydroxy-16-methyl-3,20-dioxo-pregna-1,4-dien-21-oic acid
IUPAC-namn 2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxo-acetic acid
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)C(=O)O
Specifikationer
Kemiskt namn eller material 21-Carboxy Dexamethasone
Kvantitet 50 mg
InChI formel InChI=1S/C22H27FO6/c1-11-8-15-14-5-4-12-9-13(24)6-7-19(12,2)21(14,23)16(25)10-20(15,3)22(11,29)17(26)18(27)28/h6-7,9,11,14-16,25,29H,4-5,8,10H2,1-3H3,(H,27,28)/t11-,14+,15+,16+,19+,20+,21+,22+/m1/s1
Formel vikt 406.1792