Läs mer
21-Chloro-17Alpha-[(2-furanylcarbonxyl)oxy]-9Beta,11Beta-oxido-16Alpha-methylpregna-1,4-diene-3,20-dione Mometasone Furoate Impurity D, TRC
Produktkod. 30041204
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30041204 100 mg
30040684 2.5 mg
30041123 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30041204 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC366525100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83881-09-8
Molekylformel C27 H29 Cl O6
Molekylvikt (g/mol) 484.97
Synonym (9β,11β,16α)-21-Chloro-9,11-epoxy-17-[(2-furanylcarbonyl)oxy]-16-methylpregna-1,4-diene-3,20-dione, Mometasone Furoate Imp. D (EP)
IUPAC-namn [(1S,2S,10S,11S,13R,14R,15S,17S)-14-(2-chloroacetyl)-2,13,15-trimethyl-5-oxo-18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadeca-3,6-dien-14-yl] furan-2-carboxylate
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]2(C)[C@@]1(OC(=O)c6occc6)C(=O)CCl
Specifikationer
Kemiskt namn eller material 21-Chloro-17a-[(2-furanylcarbonxyl)oxy]-9beta,11beta-oxido-16a-methylpregna-1,4-diene-3,20-dione
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C27H29ClO6/c1-15-11-19-18-7-6-16-12-17(29)8-9-24(16,2)27(18)22(33-27)13-25(19,3)26(15,21(30)14-28)34-23(31)20-5-4-10-32-20/h4-5,8-10,12,15,18-19,22H,6-7,11,13-14H2,1-3H3/t15-,18+,19+,22+,24+,25+,26+,27-/m1/s1
Formel vikt 484.1653
Procent renhet >95%