Läs mer
21-Dehydro Betamethasone (mixture of the aldehyde and the hydrated form), TRC
Produktkod. 30366429
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366429 50 mg
30365916 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30366429 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22928550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61558-12-1
Molekylformel C22 H27 F O5
Molekylvikt (g/mol) 390.45
Synonym Pregna-1,4-dien-21-al, 9-fluoro-11,17-dihydroxy-16-methyl-3,20-dioxo-, (11β,16β)-, (11β,16β)-9-Fluoro-11,17-dihydroxy-16-methyl-3,20-dioxopregna-1,4-dien-21-al, 9-Fluoro-11β,17-dihydroxy-16β-methyl-3,20-dioxopregna-1,4-dien-21-al, Betamethasone-21-aldehyde, 21-Dehydro Betamethasone, 21-Dehydrobetamethasone
IUPAC-namn 2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoacetaldehyde
LEDER C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)C=O
Specifikationer
Kemiskt namn eller material 21-Dehydro Betamethasone
Kvantitet 50 mg
InChI formel InChI=1S/C22H27FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,11-12,15-17,26,28H,4-5,8,10H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1
Formel vikt 390.1843