Läs mer
21-Dehydro Budesonide, TRC
Produktkod. 17994355
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994355 10 mg
17901485 50 mg
17912635 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17994355 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22930510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 85234-63-5
Molekylformel C25 H32 O6
Molekylvikt (g/mol) 428.52
Synonym (11β,16α)-16,17-[Butylidenebis(oxy)]-11-hydroxy-3,20-dioxo-pregna-1,4-dien-21-al, 16α,17-[(1RS)-Butylidenebis(oxy)]-11β-hydroxy-3,20-dioxopregna-1,4-dien-21-al (21-Dehydrobudesonide), Budesonide Imp. D (EP)
LEDER CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)C=O
Specifikationer
Kemiskt namn eller material 21-Dehydro Budesonide
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C25H32O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,13,16-18,20-22,28H,4-7,11-12H2,1-3H3/t16-,17-,18-,20+,21?,22+,23-,24-,25+/m0/s1
Formel vikt 428.2199
Procent renhet >95%