Läs mer
21-Dehydrocorticosterone (mixture of the aldehyde and the hydrated form), TRC
Produktkod. 30009537
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009537 250 mg
30005969 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009537 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD229160250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20287-97-2
Molekylformel C21 H28 O4
Molekylvikt (g/mol) 344.44
Synonym Pregn-4-en-21-al, 11-hydroxy-3,20-dioxo-, (11β)-, Pregn-4-en-21-al, 11β-hydroxy-3,20-dioxo- (7CI,8CI), 11β-Hydroxy-3,20-dioxo-4-pregnen-21-al, 21-Dehydrocorticosterone
IUPAC-namn 2-[(8S,9S,10R,11S,13S,14S,17S)-11-hydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]-2-oxoacetaldehyde
LEDER C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)C=O
Specifikationer
Kemiskt namn eller material 21-Dehydrocorticosterone
Kvantitet 250 mg
InChI formel InChI=1S/C21H28O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h9,11,14-17,19,24H,3-8,10H2,1-2H3/t14-,15-,16+,17-,19+,20-,21-/m0/s1
Formel vikt 344.1988