Läs mer
21-Deoxy Cortisol, TRC
Produktkod. 17983115
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17983115 5 mg
17902135 25 mg
17973425 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17983115 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2326055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 641-77-0
Molekylformel C21 H30 O4
Molekylvikt (g/mol) 346.46
Synonym Pregn-4-ene-3,20-dione, 11,17-dihydroxy-, (11β)-, Pregn-4-ene-3,20-dione, 11β,17-dihydroxy- (8CI), Progesterone, 11β,17-dihydroxy- (6CI), 11β,17α-Dihydroxypregn-4-ene-3,20-dione, 11β,17α-Dihydroxyprogesterone, 21-Dehydrohydrocortisone, 21-Deoxycortisol, 21-Deoxyhydrocortisone, 21-Desoxycortisol, Pregn-4-ene-11β,17α-diol-3,20-dione
IUPAC-namn (8S,9S,10R,11S,13S,14S,17R)-17-acetyl-11,17-dihydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
LEDER CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material 21-Deoxycortisol
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C21H30O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h10,15-18,24-25H,4-9,11H2,1-3H3/t15-,16-,17-,18+,19-,20-,21-/m0/s1
Formel vikt 346.2144
Procent renhet >95%