Läs mer
21-Deoxyprednisolone, TRC
Produktkod. 30008086
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008086 10 mg
30007545 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008086 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD24985510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20423-99-8
Molekylformel C21 H28 O4
Molekylvikt (g/mol) 344.44
Synonym Pregna-1,4-diene-3,20-dione, 11,17-dihydroxy-, (11β)-, Pregna-1,4-diene-3,20-dione, 11β,17-dihydroxy- (7CI,8CI), (11β)-11,17-Dihydroxypregna-1,4-diene-3,20-dione, 21-Deoxyprednisolone, Deprodone, Desolone, 11β,17-Dihydroxypregna-1,4-diene-3,20-dione, 21-Deoxyprednisolone
IUPAC-namn (8S,9S,10R,11S,13S,14S,17R)-17-acetyl-11,17-dihydroxy-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
LEDER CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material 21-Deoxyprednisolone
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C21H28O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h6,8,10,15-18,24-25H,4-5,7,9,11H2,1-3H3/t15-,16-,17-,18+,19-,20-,21-/m0/s1
Formel vikt 344.1988
Procent renhet >95%