Läs mer
21-Desacetyl Deflazacort, TRC
Produktkod. 30011486
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011486 25 mg
17923985 50 mg
17960845 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011486 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28869525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13649-57-5
Molekylformel C23 H29 N O5
Molekylvikt (g/mol) 399.48
Synonym 11β,21-Dihydroxy-2'-methyl-5'βH-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 5'H-Pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 11,21-dihydroxy-2'-methyl-, (11β,16β)-, 5'βH-Pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 11β,21-dihydroxy-2'-methyl- (8CI), (11β,16β)-11,21-Dihydroxy-2'-methyl-5'H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 21-Desacetyldeflazacort, Deacetyldeflazacort, Deflazacortalcohol, L 6485
IUPAC-namn (1S,2S,4R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-6,9,13-trimethyl-5-oxa-7-azapentacyclo[10.8.0.02,9.04,8.013,18]icosa-6,14,17-trien-16-one
LEDER CC1=N[C@@]2([C@@H](C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]23C)O1)C(=O)CO
Specifikationer
Kemiskt namn eller material 21-Desacetyl Deflazacort
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C23H29NO5/c1-12-24-23(18(28)11-25)19(29-12)9-16-15-5-4-13-8-14(26)6-7-21(13,2)20(15)17(27)10-22(16,23)3/h6-8,15-17,19-20,25,27H,4-5,9-11H2,1-3H3/t15-,16-,17-,19+,20+,21-,22-,23+/m0/s1
Formel vikt 399.2046
Procent renhet >95%