Läs mer
21-Desacetyldeflazacort, Mikromol™
Produktkod. 30051735
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051735 25 mg
17923826 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051735 Leverantör Mikromol Leverantörsnummer MM1224.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
21-Desacetyldeflazacort
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 21-Desacetyl Deflazacort
CAS 13649-57-5
Analyt- eller komponentnamn 21-Desacetyl Deflazacort
Frakt skick Room Temperature
Molekylformel C23 H29 N O5
Certifikat och efterlevnad ISO 17025
Synonym 21-Desacetyldeflazacort, Deacetyldeflazacort, Deflazacortalcohol, L 6485, 11beta,21-Dihydroxy-2'-methyl-5'betaH-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, (11beta,16beta)-11,21-dihydroxy-2'-methyl-5'H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 5'betaH-Pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 11beta,21-dihydroxy-2'-methyl- (8CI), (11beta,16beta)-11,21-Dihydroxy-2'-methyl-5'H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, Deflazacort Related Compound C
InChI formel InChI=1S/C23H29NO5/c1-12-24-23(18(28)11-25)19(29-12)9-16-15-5-4-13-8-14(26)6-7-21(13,2)20(15)17(27)10-22(16,23)3/h6-8,15-17,19-20,25,27H,4-5,9-11H2,1-3H3/t15-,16-,17-,19+,20+,21-,22-,23+/m0/s1
LEDER CC1=N[C@@]2([C@@H](C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]23C)O1)C(=O)CO
Molekylvikt (g/mol) 399.48
Kvantitet 25 mg
Formel vikt 399.2046 g/mol