Läs mer
21-Deschloromethyl-21-O-Ethoxycarbonyl Loteprednol Etabonate, TRC
Produktkod. 30371569
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30371569 25 mg
30375050 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30371569 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE89512025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 133991-62-5
Molekylformel C26H34O9
Molekylvikt (g/mol) 490.54
Synonym (11beta,17alpha)-17-[(Ethoxycarbonyl)oxy]-11-hydroxy-3-oxo-androsta-1,4-diene-17-carboxylic Acid Anhydride with Ethyl Hydrogen Carbonate, Prednisolone Dicarbonate
LEDER CCOC(=O)OC(=O)[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)OC(=O)OCC
Specifikationer
Kemiskt namn eller material 21-Deschloromethyl-21-O-Ethoxycarbonyl Loteprednol Etabonate
Kvantitet 25 mg
InChI formel InChI=1S/C26H34O9/c1-5-32-22(30)34-21(29)26(35-23(31)33-6-2)12-10-18-17-8-7-15-13-16(27)9-11-24(15,3)20(17)19(28)14-25(18,26)4/h9,11,13,17-20,28H,5-8,10,12,14H2,1-4H3/t17-,18-,19-,20+,24-,25-,26-/m0/s1
Formel vikt 490.2203