Läs mer
21-Hydroxy Deschloromometasone Furoate (Impurity) Mometasone Furoate Impurity H, TRC
Produktkod. 30012196
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012196 5 mg
30009845 25 mg
30003374 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012196 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9487305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 148596-90-1
Molekylformel C27 H31 Cl O7
Molekylvikt (g/mol) 502.98
Synonym (11β,16α)-9-Chloro-17-[(2-furanylcarbonyl)oxy]-11,21-dihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9-chloro-11β,21-dihydroxy-16α-methyl-3,20-dioxopregna-1,4-dien-17-yl furan-2-carboxylate, Mometasone Furoate Imp. H (EP)
IUPAC-namn [(8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)CO
Specifikationer
Kemiskt namn eller material 9-Chloro-11beta,21-dihydroxy-16alpha-methyl-3,20-dioxopregna-1,4-dien-17-yl Furan-2-carboxylate
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C27H31ClO7/c1-15-11-19-18-7-6-16-12-17(30)8-9-24(16,2)26(18,28)21(31)13-25(19,3)27(15,22(32)14-29)35-23(33)20-5-4-10-34-20/h4-5,8-10,12,15,18-19,21,29,31H,6-7,11,13-14H2,1-3H3/t15-,18+,19+,21+,24+,25+,26+,27+/m1/s1
Formel vikt 502.1758
Procent renhet >95%