Läs mer
(22S)-Budesonide, TRC
Produktkod. 30133107
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133107 25 mg
30133976 10 mg
30007963 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133107 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68950525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51372-28-2
Molekylformel C25H34O6
Molekylvikt (g/mol) 430.53
Synonym (11beta,16alpha)-16,17-[(1S)-Butylidenebis(oxy)]-11,21-dihydroxypregna-1,4-diene-3,20-dione, Budesonide Epimer A (USP)
IUPAC-namn (1S,2S,4R,6S,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-16-one
LEDER CCC[C@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Specifikationer
Kemiskt namn eller material (22S)-Budesonide
Kvantitet 25 mg
InChI formel InChI=1S/C25H34O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-18,20-22,26,28H,4-7,11-13H2,1-3H3/t16-,17-,18-,20+,21-,22+,23-,24-,25+/m0/s1
Formel vikt 430.2355