Läs mer
24,25-Dihydroxy Fusidic Acid, TRC
Produktkod. 30012402
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012402 100 mg
30008360 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012402 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD452840100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80445-74-5
Molekylformel C31 H50 O8
Molekylvikt (g/mol) 550.7239
Synonym 29-Nordammar-17(20)-en-21-oic acid, 16-(acetyloxy)-3,11,24,25-tetrahydroxy-, (3α,4α,8α,9β,11α,13α,14β,16β,17Z)- (9CI), 24,25-Dihydroxyfusidic acid, ent-(24SR,17Z)-16α-(Acetyloxy)-3β,11β,24,25-tetrahydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholest-17(20)-en-21-oic acid, 24,25-Dihydro-24,25-dihydroxyfusidic acid
IUPAC-namn (2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-5,6-dihydroxy-6-methylheptanoic acid
LEDER C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC(O)C(C)(C)O)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C
Specifikationer
Kemiskt namn eller material 24,25-Dihydroxy Fusidic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C31H50O8/c1-16-19-10-13-30(6)26(29(19,5)12-11-21(16)33)22(34)14-20-25(23(39-17(2)32)15-31(20,30)7)18(27(36)37)8-9-24(35)28(3,4)38/h16,19-24,26,33-35,38H,8-15H2,1-7H3,(H,36,37)/b25-18-/t16-,19-,20-,21+,22+,23-,24?,26-,29-,30-,31-/m0/s1
Formel vikt 550.3506