missing translation for 'onlineSavingsMsg'
Läs mer

24-epi-Castasterone, TRC

Product Code. 30363621
Change view
Click to view available options
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30363621 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363621 Supplier Toronto Research Chemicals Supplier No. TRCC2264901MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 24-epi-Castasterone
CAS 72050-71-6
Molekylformel C28 H48 O5
Kvantitet 1 mg
Synonym Ergostan-6-one, 2,3,22,23-tetrahydroxy-, (2α,3α,5α,22R,23R)-, (2α,3α,5α,22R,23R)-2,3,22,23-Tetrahydroxyergostan-6-one, 24-Epicastasterone, 24-epi-Castasterone, 24R-Epicastasterone
InChI formel InChI=1S/C28H48O5/c1-14(2)15(3)25(32)26(33)16(4)18-7-8-19-17-11-22(29)21-12-23(30)24(31)13-28(21,6)20(17)9-10-27(18,19)5/h14-21,23-26,30-33H,7-13H2,1-6H3/t15-,16+,17+,18-,19+,20+,21-,23+,24-,25-,26-,27-,28-/m1/s1
LEDER CC(C)[C@@H](C)[C@@H](O)[C@H](O)[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC(=O)[C@H]4C[C@H](O)[C@H](O)C[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn (2R,3S,5S,8S,9S,10R,13S,14S,17R)-17-[(2S,3R,4R,5R)-3,4-dihydroxy-5,6-dimethylheptan-2-yl]-2,3-dihydroxy-10,13-dimethyl-1,2,3,4,5,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-6-one
Molekylvikt (g/mol) 464.678
Formel vikt 464.35
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.