Läs mer
24-Hydroxycholesterol, MedChemExpress
Produktkod. 16451718
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
Förpackningsstorlek:
2mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16451718 5 mg 5mg
16440396 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16451718 Leverantör Medchem Express Leverantörsnummer HYN2370
MedChemExpress 24-Hydroxycholesterol is a natural sterol, which serves as a positive allosteric modulator of N-Methyl-d-Aspartate (NMDA) receptorsR, and a potent activator of the transcription factors LXR.
24-Hydroxycholesterol is a natural sterol, which serves as a positive allosteric modulator of N-Methyl-d-Aspartate (NMDA) receptorsR, and a potent activator of the transcription factors LXR.
Specifikationer
Kemiskt namn eller material 24-Hydroxycholesterol
CAS 30271-38-6
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C27H46O2
Kvantitet 5 mg
Löslighetsinformation DMSO : 25 mg/mL (62.09 mM; Need ultrasonic)
LEDER CC(C)C(O)CC[C@@H](C)[C@H]1CC[C@@]2([H])[C@]3([H])CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C
Molekylvikt (g/mol) 402.65
Formel vikt 402.65
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neurodegeneration