Läs mer
24-Hydroxycholesterol, TRC
Produktkod. 30012051
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012051 10 mg
30005192 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012051 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH91804010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 30271-38-6
Molekylformel C27H46O2
Molekylvikt (g/mol) 402.65
Synonym Cholest-5-ene-3β,24-diol, (3β)-Cholest-5-ene-3,24-diol
IUPAC-namn (3S,8S,9S,10R,13R,14S,17R)-17-[(2R)-5-hydroxy-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER CC(C)C(O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 24-Hydroxycholesterol
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C27H46O2/c1-17(2)25(29)11-6-18(3)22-9-10-23-21-8-7-19-16-20(28)12-14-26(19,4)24(21)13-15-27(22,23)5/h7,17-18,20-25,28-29H,6,8-16H2,1-5H3/t18-,20+,21+,22-,23+,24+,25?,26+,27-/m1/s1
Formel vikt 402.3498
Procent renhet >95%