Läs mer
25-Desacetyl Rifapentin, TRC
Produktkod. 30368661
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30368661 10 mg
1 options
Produktkod. 30368661 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28872010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 25-Desacetyl Rifapentin
CAS 79039-56-8
Molekylformel C45 H62 N4 O11
Kvantitet 10 mg
Synonym 25-Desacetylrifapentine, Rifamycin, 3-[[(4-cyclopentyl-1-piperazinyl)imino]methyl]-25-O-deacetyl-, 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan, rifamycin deriv., 3-[[(4-Cyclopentyl-1-piperazinyl)imino]methyl]-25-O-deacetylrifamycin, 25-Deacetylrifapentine, 25-Desacetyl rifapentin, L 14583
InChI formel InChI=1S/C45H62N4O11/c1-23-12-11-13-24(2)44(57)47-35-30(22-46-49-19-17-48(18-20-49)29-14-9-10-15-29)40(54)32-33(41(35)55)39(53)28(6)42-34(32)43(56)45(7,60-42)59-21-16-31(58-8)25(3)37(51)27(5)38(52)26(4)36(23)50/h11-13,16,21-23,25-27,29,31,36-38,50-55H,9-10,14-15,17-20H2,1-8H3,(H,47,57)/b12-11+,21-16+,24-13-,46-22+/t23-,25+,26+,27-,31-,36-,37+,38+,45?/m0/s1
LEDER CO[C@H]1\C=C\OC2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](O)[C@@H]1C)\C)c(\C=N\N5CCN(CC5)C6CCCC6)c(O)c4c3C2=O
Molekylvikt (g/mol) 834.99
Formel vikt 834.4415